Adhesive qPCR film seals, QuickSeal qPCR Crystal™
Clear, non-tacky, pressure sensitive film for qPCR plates (96- or 384-well) and situations where fluorescence is experienced.
- Optically clear, minimal auto-fluorescence
- DMSO resistant
- Non-pierceable, peelable
- Seal integrity range: −40 to +110 °C
- Available sterile or non sterile
Frequently Bought Together