Order Entry
Belgium
Orders LinkContactUsLinkComponent
Proteinase K solution

Proteinase K solution

Proteinase K solution
Supplier:  PanReac AppliChem
Certificates
Proteinase K solution

3 Options Available Below

Product Details & Documents

Proteinase K is used to destruct proteins in cell lysates (tissue, cultured cells) and to liberate nucleic acids, since it very effectively digests DNases and RNases.

  • DNases/RNases not detectable
  • Proteinase K min. 600 mAnsonU/ml
  • GHS symbols: GHS08
  • GHS signal word: Danger

Composition:
CaCl₂ · 2H₂O 1 mM
Glycerol (v/v) 40%
Proteinase K 20 mg/ml
Tris · HCl (pH 7,5) 10 mM

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • A4392.0001
  • A4392.0005
  • A4392.0010
  • Enzyme name:
  • Proteinase K solution
  • Proteinase K solution
  • Proteinase K solution
  • Enzyme synonyms:
  • Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
  • Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
  • Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
  • Concentration:
  • 20 mg/ml
  • 20 mg/ml
  • 20 mg/ml
  • Storage conditions:
  • 2...8 °C
  • 2...8 °C
  • 2...8 °C

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 46,60 €
    46,60 €
    1 * 1 mL
       46,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    A4392.0001
    Enzyme name
    Proteinase K solution
    Enzyme synonyms
    Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
    Concentration
    20 mg/ml
    Storage conditions
    2...8 °C
  • 5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 126,00 €
    126,00 €
    1 * 5 mL
       126,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    A4392.0005
    Enzyme name
    Proteinase K solution
    Enzyme synonyms
    Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
    Concentration
    20 mg/ml
    Storage conditions
    2...8 °C
  • 10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 226,00 €
    226,00 €
    1 * 10 mL
       226,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    A4392.0010
    Enzyme name
    Proteinase K solution
    Enzyme synonyms
    Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
    Concentration
    20 mg/ml
    Storage conditions
    2...8 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.