Order Entry
Belgium
ContactUsLinkComponent
Mag-Bind® RXNPure Plus kit
Mag-Bind® RXNPure Plus kit
Catalog # M1386-01
Supplier:  OMEGA BIO-TEK
CAS Number:  
Mag-Bind® RXNPure Plus kit
Catalog # M1386-01
Supplier:  OMEGA BIO-TEK
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® RXNPure Plus Kit
  • Sample size:
    Variable
  • Description:
    Mag-Bind® RXNPure Plus Kit

Specifications

About this item

Mag-Bind® RXNPure Plus Kit allows rapid and reliable isolation DNA from PCR and enzymatic reactions with high recovery rates. The system combines proprietary chemistries with the reversible nucleic acid-binding properties of magnetic beads that selectively bind PCR amplicons 100 bp and larger, and eliminate excess nucleotides, primers and small, non targeted amplification products, such as primer dimers.

  • Efficiently removes excess primers, primer-dimers, dNTPs and salts
  • No centrifugation/filtration steps
  • Scalable - can be adapted to most standard liquid handling robots
  • Use in 96- or 384- well format

This kit is designed for both manual and fully automated purification and may not require reprogramming of liquid handling instruments depending on your current method.