Spectroscopy cuvettes
These high quality cuvettes allows you to make full use of the accuracy and sensitivity of your optical instrument.
- High transmission
- Accurate pathlength
- Windows are fused to the bodies to avoid any deformation
These high quality cuvettes allows you to make full use of the accuracy and sensitivity of your optical instrument.
- High transmission
- Accurate pathlength
- Windows are fused to the bodies to avoid any deformation
Product Family Options
- DescriptionTypeBase thicknessVolumePath lengthWindow material
- Cuvettes with PTFE lidMicro3,2 mm700 µl10 mmOptical glass1 * 2 ST202,00 €
- Cuvettes with PTFE lidMicro3,2 mm700 µl10 mmQuartz (Suprasil®)1 * 2 ST1 310,00 €