Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Flammable
Irritant
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Catalog # 85022.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Catalog # 85022.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Residue on evaporation:
    Max. 0.0003 %
  • Water:
    Max. 0.0050 %
  • Stabilizer (EtOH):
    0.5 - 1.5 % w/v%
  • —:

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
ADR:3,I
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :Stabilised with ethanol 0.5 - 1.5 % (w/v)
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646