Orders LinkContactUsLinkComponent
Counting machine
Counting machine
Counting machine, 5-place
Catalog # 720-2761
Supplier:   HAWKSLEY AND SONS
Counting machine
Catalog # 720-2761
Supplier:  HAWKSLEY AND SONS
Counting machine
Counting machine, 5-place
Price
670,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Totaliser five place counting machine. An intuitive differential cell counter that simplifies blood cell counts.
  • When a total of 100 cells is counted, a bell rings
  • Percentage of each type of cell can then be read directly from the window above each key
  • simple counter-clockwise rotation of the end knobs will zero all the counts
  • Each key has the name and picture of a particular cell above it, representing groups of leucocytes
  • The key legend strips are easily removed to allow customisation
  • Five place set to count Basophils, Eosinophils, Seg., Lymphocytes, Monocytes
  • Sturdy solid construction with firm dependable keys
Specifications
  • Description:
    Counting machine, 5-place