Order Entry
Belgium
ContactUsLinkComponent
Surface cleaner for use in ultrasonic baths, STAMMOPUR R
Surface cleaner for use in ultrasonic baths, STAMMOPUR R
Catalog # 142-1904
undefined
Surface cleaner for use in ultrasonic baths, STAMMOPUR R
Catalog # 142-1904

Specifications

  • Description:
    STAMMOPUR R cleaning concentrate
  • Packed:
    5 litre PE container
  • Pk:
    5 L
  • pH value:
    9,6 (1%)

Specifications

About this item

STAMMOPUR R is a mildly alkaline, cleaning concentrate for cleaning of medical instruments and devices in an ultrosonic bath.

  • High cleaning efficiency and blood dissolution
  • Pleasant odour
  • Contains propan-2-ol (<10%), surfactants, complexing agents, alkali
  • Very high material compatibility