ContactUsLinkComponent
Swing over bench
Swing over bench
  134-0501
 :  Palbam
undefined
Swing over bench
  134-0501
 :  Palbam
  • Description:
    Swing over bench
  • W×D×H:
    850×350×450 mm
  • Pk:
    1
This bench is specifically designed to assist in putting on over shoes - by supporting the foot with a small raised platform. Two or more benches can be connected together (either seat-to-seat or foot support-to-foot support) to create the ideal step over bench.
  • The bench can also be used as a separation barrier between pre-gowning and gowning areas
  • Optional connection pieces allow benches to be joined together (see accessories)
Stainless steel 304 with electropolished finish.
 : Typically available from stock for direct shipment from the supplier.