Dry block thermostat for cuvettes, LT 200
: Hach
A digestion step often has to be carried out before key parameters can be determined. For these parameters, the LT 200 thermostat complements the LANGE cuvette test system. Two separately controllable heating blocks enable cuvettes and reaction vessels to be digested at identical or different temperatures and time settings. The LT 200 has a digital timer with an automatic switch-off and acoustic signal. Two transparent splash protection lids cover the thermostat while it is heating. The integrated over-temperature feature and the insulated external casing provide additional safety.
- Flexible as pre-programmed for all standard digestions and freely programmable for user specific digestions
- Pre-programmed for 40, 100 and 148 °C. User specific freely selectable programs from 37 to 148 °C for 1 to 480 minutes
- Heating rate is from 20 to 148 °C in 10 minutes
- Excellent reproducibility due to very good temperature stability
- Simple to use with large, easily readable display and one-key operation for standard digestions
: Supplied with different block configurations, please see below.
Frequently Bought Together