Orders LinkContactUsLinkComponent
Probes for temperature loggers EBI 300 and 310 (Ebro)
Probes for temperature loggers EBI 300 and 310 (Ebro)
Supplier:   EBRO
Probes for temperature loggers EBI 300 and 310 (Ebro)
Supplier:  EBRO
Certificates
Probes for temperature loggers EBI 300 and 310 (Ebro)
Probes for temperature loggers EBI 300 and 310 (Ebro)
10 Options Available Below
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature.
Product Family Options
Product Information
  • Type
    Range
    Accuracy
    Availability
    Price
  • 620-1860
    External replacement probe for EBI 300 TE, Ø×L: 4×50 mm, TPC 300
    External replacement probe for EBI 300 TE, Ø×L: 4×50 mm, TPC 300
    TPC 300 (NTC)
    –35...+75 °C
    T: ±0,5 °C (–20…+40 °C); ±0,8 °C for the remaining measurement range
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,00 €
    77,00 €
    1 * 1 ST
       77,00 €
  • 620-1861
    External replacement probe for EBI 310 TE, Ø×L: 5×50 mm, TPX 220
    External replacement probe for EBI 310 TE, Ø×L: 5×50 mm, TPX 220
    −200...+250 °C
    T: ±2,0 °C (–200…–100 °C); ±1,0 °C (–100 …–20 °C); ±0,2 °C (–20…+60 °C); ±0,5 °C (+60…+250 °C)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • 620-2394
    External replacement probe for EBI 310TH, TPH 500
    External replacement probe for EBI 310TH, TPH 500
    –30… +75 °C; 0 - 100 % RH
    RH: ±2% between 10 - 90% (at +25 °C); ±4 % for the remaining measuring range
    T: ±0,5 °C (+20… +60 °C); ±0,8 °C for the remaining measuring range
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • 620-2762
    External replacement probe for EBI 310TX- P1, Ø×L: 5×45 mm, blunt, with 3 m PTFE cable
    External replacement probe for EBI 310TX- P1, Ø×L: 5×45 mm, blunt, with 3 m PTFE cable
    −200...+200 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 620-2763
    External replacement probe for EBI 310TX- P2, Ø×L: 5×45 mm, blunt, with 3 m PTFE cable
    External replacement probe for EBI 310TX- P2, Ø×L: 5×45 mm, blunt, with 3 m PTFE cable
    −50...+180 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 620-2764
    External replacement probe for EBI 310TX- P3, Ø×L: 5×45 mm, blunt, with 1 m PTFE cable
    External replacement probe for EBI 310TX- P3, Ø×L: 5×45 mm, blunt, with 1 m PTFE cable
    −50...+180 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 620-2765
    External replacement probe for EBI 310TX- P4, Ø×L: 5×45 mm, blunt, with 3 m metal wrapped cable
    External replacement probe for EBI 310TX- P4, Ø×L: 5×45 mm, blunt, with 3 m metal wrapped cable
    100...400 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 371,00 €
    371,00 €
    1 * 1 ST
       371,00 €
  • 620-2823
    External replacement probe for PT 1000, Ø×L: 5×50 mm, TPX 220-3
    External replacement probe for PT 1000, Ø×L: 5×50 mm, TPX 220-3
    −200...+250 °C
    T: ±2,0 °C (–200…–100 °C); ±1,0 °C (–100 …–20 °C); ±0,2 °C (–20…+60 °C); ±0,5 °C (+60…+250 °C)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • 620-2262
    Internal replacement probe for EBI 300 TH, TPH 400
    Internal replacement probe for EBI 300 TH, TPH 400
    TPH 300 (NTC, capacitive)
    –30… +70 °C; 0 - 100 % RH
    RH: ±3% between 10 - 90% (at +25 °C); ±5 % for the remaining measuring range
    T: ±0,5 °C (+20… +40 °C); ±1 °C for the remaining measuring range
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • 620-2761
    Replacement adapter for EBI 310 TX, type TPX 310
    Replacement adapter for EBI 310 TX, type TPX 310
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 174,00 €
    174,00 €
    1 * 1 ST
       174,00 €