Order Entry
Belgium
Orders LinkContactUsLinkComponent
Label, removable

Label, removable

Removable, square labels for all H20 chip tray covers, H-20, Polyolefin, Repositionable, rubber based, White, 38×38 mm, 1000 labels/roll
Catalog # ENTEH20-002LB
Supplier:   ENTEGRIS
Label, removable
Catalog # ENTEH20-002LB
Supplier:  ENTEGRIS
Label, removable
Label, removable

Removable, square labels for all H20 chip tray covers, H-20, Polyolefin, Repositionable, rubber based, White, 38×38 mm, 1000 labels/roll

Price
Inquire For Price
Log in for availability
1 * 1 ROU
Product Details & Documents

Made of polyolefin copolymer. Blank label for customized marking. Pressure-sensitive adhesive is both removable and nondepositing.

  • Adheres well to most plastic surfaces
  • For use with all H20 chip tray covers
  • Leaving no adhesive residue
Specifications
  • Type:
    H-20
  • Colour:
    White
  • Description:
    Removable, square labels for all H20 chip tray covers
  • Dimensions:
    38×38 mm
  • Material:
    Polyolefin
  • Packed:
    1000 labels/roll
  • Adhesive:
    Repositionable, rubber based