Orders LinkContactUsLinkComponent
Sample dipper with v bottom
Sample dipper with v bottom
Supplier:   ENTEGRIS
Sample dipper with v bottom
Supplier:  ENTEGRIS
Certificates
Sample dipper with v bottom
3 Options Available Below
Product Details & Documents
Holds a wafer or a square plate.
  • Slot has 'V' bottom
  • Clip on handle slides down to hold substrates in place
  • Hold one wafer or one square plate
Documents
Product Family Options
Product Information
  • Availability
    Price
  • ENTED11-0215
    Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
    Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST
  • ENTED14-0215
    Dipper, wafer size 57 - 125 mm, square plate size 50 - 153 mm, max. thickness 3,0 mm
    Dipper, wafer size 57 - 125 mm, square plate size 50 - 153 mm, max. thickness 3,0 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST
  • ENTED15-0215
    Dipper, wafer size 125 - 200 mm, square plate size 114 - 203 mm, max. thickness 6,4 mm
    Dipper, wafer size 125 - 200 mm, square plate size 114 - 203 mm, max. thickness 6,4 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST