ContactUsLinkComponent
Analytical HPLC columns, XSelect™ CSH C18
Analytical HPLC columns, XSelect™ CSH C18
Supplier:  WATERS
Certificates
Analytical HPLC columns, XSelect™ CSH C18
Supplier:  WATERS
Specifications
  • Catalog No:
  • WATE186005262
  • WATE186005270
  • Carbon load:
  • 15%
  • 15%
  • Endcapped:
  • Yes
  • Yes
  • Surface area:
  • 185 m²/g
  • 185 m²/g
  • USP code:
  • L1
  • L1