Sampling scoops, Behrotest®
Supplier: BEHR
Jump to:
About this item
The sampling scoop is composed of a clamping fixture and end bar. The scoop with receptacles made of borosilicate glass comply with the high requirements.
- Sturdy three-part supporting bar with telescopic racks
- Continuously adjustable in length
Suitable for sampling in waters that are difficult to access, from the surface.
: EN 1485/AOX
3 Options Available Below
Product Details & Documents
The sampling scoop is composed of a clamping fixture and end bar. The scoop with receptacles made of borosilicate glass comply with the high requirements.
- Sturdy three-part supporting bar with telescopic racks
- Continuously adjustable in length
Suitable for sampling in waters that are difficult to access, from the surface.
: EN 1485/AOX
Product Family Options
Product Information
- ModelCapacityLengthMaterialAvailabilityPrice
- BEHRB00196081Sampling scoop with beakerSampling scoop with beakerPP 10001000 ml1 − 3,75 mStainless steel/PPLog in for availability1 * 1 ST500,00 €
- 331-3110Sampling scoop with borosilicate glass beakerSampling scoop with borosilicate glass beaker1000 ml1 − 3,75 mStainless steelLog in for availability1 * 1 ST506,00 €
- 331-3120Sampling scoop with high breast bottleSampling scoop with high breast bottle1000 ml1 − 3,75 mStainless steelLog in for availability1 * 1 ST509,00 €