ContactUsLinkComponent
VWR® WebSeal™, Microplate Sealing Mats
VWR® WebSeal™, Microplate Sealing Mats
  548-3395
undefined
VWR® WebSeal™, Microplate Sealing Mats
  548-3395
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Sealmat (block cover), 96 round well shape, Ø 8 mm, alcohol resistant
  • Material:
    EVA
  • Seq:
    05
  • Pk:
    5