ContactUsLinkComponent
Single-use data loggers, Testo 184 T1 and Testo 184 T2
Single-use data loggers, Testo 184 T1 and Testo 184 T2
Supplier:  TESTO
Certificates
undefined
Single-use data loggers, Testo 184 T1 and Testo 184 T2
Supplier:  TESTO
Specifications
  • Catalog No:
  • 620-2309
  • 620-2306
  • Sensor:
  • Internal NTC sensor
  • Internal NTC sensor
  • Alarm:
  • Visual by LEDs
  • Visual by LEDs
  • Protection class:
  • IP 67
  • IP 67
  • Recording intervals:
  • 1 min. - 24 h
  • 1 min. - 24 h
  • Weight:
  • 44 g
  • 22 g
  • W×D×H:
  • 44×12×97 mm
  • 35×9×75 mm
  • Data memory - No. of readings:
  • 40000
  • 16000
  • Batteries:
  • Non-replaceable, lithium
  • Non-replaceable, lithium