Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Mycotoxin
Solid phase extraction cartridges, Bond Elut™ Mycotoxin
Bond Elut™ Jr, Mycotoxin, 500 mg
Catalog # VARI12165001B
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Mycotoxin
Catalog # VARI12165001B
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ Mycotoxin
Bond Elut™ Jr, Mycotoxin, 500 mg
Price
655,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Bond Elut™ Mycotoxin is a novel sorbent which cleans up food extracts for improved trichothecene and zearalenone analysis. Results are comparable or superior to competing methods, including immunoaffinity columns (IaC) and charcoal/alumina columns. The sorbent is a proprietary silica-based ion exchange material.
  • Simple methodology saves time and increases throughput
  • Use with a broad range of food matrices
  • Alternative to immunoaffinity techniques
The Bond Elut™ Mycotoxin method for extraction and cleanup is successful with a variety of food and grain sample types, including wheat, corn, durum, oats, bread, muesli and infant food.
Bond Elut™ Mycotoxin is easy to use and acts in a selective non-retention way – the toxin analytes pass through the cartridge while the food matrix components are retained.
Specifications
  • Description:
    Bond Elut™ Jr
  • Bed weight:
    500 mg
  • Phase:
    Mycotoxin