ContactUsLinkComponent
Cotton roving for pipette plugger
Cotton roving for pipette plugger
Catalog # BELC7750-22222
Supplier:  BELLCO GLASS
undefined
Cotton roving for pipette plugger
Catalog # BELC7750-22222
Supplier:  BELLCO GLASS
Specifications
  • Description:
    Absorbent cotton roving for pipette plugging
  • Pk:
    1 kg
Specifications
About this item
Cotton roving made of 12 grain premium long fiber cotton.
Sufficient for approx. 35000 pipettes.
Packaging: Coiled in plastic bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904