Order Entry
Belgium
ContactUsLinkComponent
Tape dispenser, single roll
Tape dispenser, single roll
Catalog # 817-0309
Supplier:  ULTRATAPE
undefined
Tape dispenser, single roll
Catalog # 817-0309
Supplier:  ULTRATAPE

Specifications

  • For Use With:
    Tapes with 51 mm width
  • Colour:
    Blue
  • Description:
    Cleanroom tape dispenser, single roll
  • Pk:
    1

Specifications

About this item

UltraTape single roll dispenser is designed to hold one roll up to 50,8 mm width and 201,2 metres in length. The dispenser is supplied with a 76 mm plastic core holder.