Urine collection system, UriSwab
Catalog # SLIN800C
Supplier: STERILIN
Specifications
- Description:Urine collection systems
- Environmentally Preferable:
- Pk:500
Specifications
About this item
A unique sponge swab is impregnated with the exact amount of boric acid for the sample collected and hence no fill line is required.
- Ideal for use in automation
- Simplifies collection of urine samples
Suitable for the collection, transport, and preservation of urine samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904