Orders LinkContactUsLinkComponent
Self-laminating labels, MiniMark
Self-laminating labels, MiniMark
Self-laminating labels, MNK-BPT-211-427, Vinyl, White/clear, 25×25 mm
Catalog # BRDY710113
Supplier:   Brady
Self-laminating labels, MiniMark
Catalog # BRDY710113
Supplier:  Brady
Self-laminating labels, MiniMark
Self-laminating labels, MiniMark
Self-laminating labels, MNK-BPT-211-427, Vinyl, White/clear, 25×25 mm
Price
444,00 €
Log in for availability
1 * 1 ROU
Product Details & Documents
The MiniMark self-laminating wire and cable markers are designed to adhere to highly curved surfaces, wires and cables. They feature a white printable area with clear tail that wraps around marker. The clear tail protects the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling.
Specifications
  • Colour:
    White/clear
  • Description:
    Self-laminating labels
  • Dimensions:
    25×25 mm
  • Material:
    Vinyl
  • Model:
    MNK-BPT-211-427