Orders LinkContactUsLinkComponent
Pipette tips
Supplier:   Alpha Laboratories
Pipette tips
Supplier:  Alpha Laboratories
Certificates
Pipette tips
11 Options Available Below
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
  • Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
  • Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
  • With graduations to visually check volume
  • Suitable for all major pipettor brands
Product Family Options
Product Information
  • Tip style
    Volume
    Sterile
    Length
    Colour code
    Packed
    Availability
    Price
  • Micro, ultrafine
    0,1 - 10 µl
    -
    32 mm
    Natural
    96 tips/rack; 12 racks/case
    Log in for availability
    Log in for availability
    1 * 1.152 ST
    1 * 1.152 ST - 176,00 €
    176,00 €
    1 * 1.152 ST
       176,00 €
  • Micro, ultrafine
    0,1 - 10 µl
    +
    32 mm
    Natural
    96 tips/rack; 12 racks/case
    Log in for availability
    Log in for availability
    1 * 1.152 ST
    1 * 1.152 ST - 198,00 €
    198,00 €
    1 * 1.152 ST
       198,00 €
  • Micro, ultrafine
    0,1 - 10 µl
    -
    32 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,10 €
    54,10 €
    1 * 1.000 ST
       54,10 €
  • Bevelled, non-graduated
    1 - 200 µl
    -
    50 mm
    Yellow
    96 tips/rack; 10 racks/case
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 145,00 €
    145,00 €
    1 * 960 ST
       145,00 €
  • Bevelled, non-graduated
    1 - 200 µl
    +
    50 mm
    Yellow
    96 tips/rack; 10 racks/case
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 167,00 €
    167,00 €
    1 * 960 ST
       167,00 €
  • Bevelled, non-graduated
    1 - 200 µl
    -
    50 mm
    Yellow
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,10 €
    54,10 €
    1 * 1.000 ST
       54,10 €
  • Bevelled, graduated
    1 - 200 µl
    -
    50 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,10 €
    54,10 €
    1 * 1.000 ST
       54,10 €
  • Bevelled, graduated
    1 - 200 µl
    +
    50 mm
    Natural
    96 tips/rack; 10 racks/case
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 177,00 €
    177,00 €
    1 * 960 ST
       177,00 €
  • Bevelled, non-graduated
    200 - 1000 µl
    -
    76 mm
    Blue
    96 tips/rack; 6 racks/case
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 127,00 €
    127,00 €
    1 * 576 ST
       127,00 €
  • Bevelled, non-graduated
    200 - 1000 µl
    -
    76 mm
    Blue
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,60 €
    90,60 €
    1 * 1.000 ST
       90,60 €
  • Bevelled, non-graduated
    200 - 1000 µl
    +
    76 mm
    Blue
    96 tips/rack; 6 racks/case
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 144,00 €
    144,00 €
    1 * 576 ST
       144,00 €