Orders LinkContactUsLinkComponent
Woulff bottles
Woulff bottles
500 ml, with two side necks GL 18, center neck GL 25, complete with screw caps
Catalog # 519-4500
Supplier:   Sartorius
Woulff bottles
Catalog # 519-4500
Supplier:  Sartorius
Woulff bottles
Woulff bottles
500 ml, with two side necks GL 18, center neck GL 25, complete with screw caps
Price
167,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of borosilicate glass.
  • Side necks with screw caps and inlet/outlet glass tubes
  • Centre neck with screw cap and stopcock
  • Used between suction flask and vacuum source
  • Very good chemical and temperature resistance
With three necks. For protection of vacuum pump.
Specifications
  • Capacity:
    500 ml
  • Neck:
    with two side necks GL 18, center neck GL 25, complete with screw caps
  • Cat. No.:
    519-4500
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Tap, glass
    Tap, glass
    SART6980000-37
    Woulff's bottles
    Log in for availability
    1 * 1 ST - 29,50 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.