Orders LinkContactUsLinkComponent
Detergent, Diversey Attack Plus E9e
Detergent, Diversey Attack Plus E9e
Diversey Attack Plus E9e, detergent, multi-purpose, 2×5 l containers/case
Catalog # DIVY7516886
Supplier:   Diversey
Detergent, Diversey Attack Plus E9e
Catalog # DIVY7516886
Supplier:  Diversey
Detergent, Diversey Attack Plus E9e
Diversey Attack Plus E9e, detergent, multi-purpose, 2×5 l containers/case
Price
91,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Clear yellow, heavy-duty cleaner containing a mixture of organic solvents and compounds.
  • Good emulsifying formula
  • Removes grease and dirt
  • Medium foaming alkaline formula, pH of neat solution: 12
Specifications
  • Description:
    Diversey Attack Plus E9e, detergent, multi-purpose
  • Seq:
    01
  • Packed:
    2×5 l containers/case