ContactUsLinkComponent
Disinfectant detergent, Delladet VS2
Disinfectant detergent, Delladet VS2
  DIVY7509345
 :  Diversey
undefined
Disinfectant detergent, Delladet VS2
  DIVY7509345
 :  Diversey
  • Description:
    Delladet VS2, disinfectant detergent
  • Seq:
    01
  • Packed:
    5 litre container
  • Pk:
    2 Can
A concentrated detergent with disinfectant properties, specially formulated for open plant cleaning applications in the food, beverage and dairy industries. Delladet is recommended for cleaning and disinfection phase of floors, walls, utensils, equipment and other surfaces in contact with foodstuffs.
  • Contains a blend of high foaming surfactant, sequestrant and alkaline builders
  • Broad range disinfecting properties are provided by the incorporation of quaternary ammonium compound (QAC)
  • Suitable for use in soft or hard water
  • Rinse thoroughly with potable water after application
 : Bactericide EN 1276.
 : Use biocides safely. Always read the label and product information before use.