Overalls, Primeguard, PP
Supplier: Synergy Health
Certificates
About this item
These disposable economy overalls are made of PP. They are designed for protection against airborne particles and limited non toxic splash and spray. Ideal for visitors.
- Hood
- Elasticated wrists and ankles
- Anti-static
Colour: White or blue
Weight: 50 g/m²
Type 5/6
Specifications
- Catalog No:
- VEROPG5X11
- VEROPG5XX11
- Model:
- Primeguard
- Primeguard
- Anti-static:
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Weight:
- 50 g/m²
- 50 g/m²