ContactUsLinkComponent
Surface cleaning and disinfectant, terralin® PAA
Surface cleaning and disinfectant, terralin® PAA
  SCUE126203
undefined
Surface cleaning and disinfectant, terralin® PAA
  SCUE126203
  • Description:
    terralin® PAA, surface disinfectant
  • Packed:
    2 × 80 ml bottles (12 double bottles per carton)
  • Pk:
    12
A two component formulation that generates active oxygen for cleaning and disinfection of surfaces and medical devices. Applications include targeted wiping disinfection and cleaning e.g. in the case of an outbreak.
  • Effective against bacteria, fungi, viruses and spores (incl. Clostridium) tuberculocidal, mycobactericidal
  • Very short contact time
  • Effective immediately upon preparation of 2% solution - typically add the contents of both bottles into 4 litres of water
 : Bactericidal EN 13727; Fungicidal EN 13624; Mycobactericidal EN 14348; Sporicidal EN 14347; Virucidal EN 14476.