

2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:200-661-7
:1219
:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:12 °C
:01-2119457558-25
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Assay (on anhydrous substance):≥ 99,9 %
- Acidity:≤ 0,0002 meq/g
- Alkalinity:≤ 0,0002 meq/g
- Evaporation residue:≤ 1 ppm
- Water:≤ 50 ppm
- Ag (Silver):≤ 10 ppb
- Al (Aluminium):≤ 25 ppb
- Ca (Calcium):≤ 25 ppb
- Cu (Copper):≤ 10 ppb
- Fe (Iron):≤ 10 ppb
- K (Potassium):≤ 10 ppb
- Mg (Magnesium):≤ 10 ppb
- Mn (Manganese):≤ 10 ppb
- Na (Sodium):≤ 50 ppb
- Ni (Nickel):≤ 10 ppb
- Pb (Lead):≤ 10 ppb
- Zn (Zinc):≤ 10 ppb
- Transmittance (205 nm):≥ 10 %
- Transmittance (210 nm):≥ 50 %
- Transmittance (225 nm):≥ 80 %
- Transmittance (240 nm):≥ 95 %
- Transmittance (255 nm):≥ 99 %
- Suitable for LC-MS:Passes test
Frequently Bought Together