Orders LinkContactUsLinkComponent
Mat frames for PurusMat
Mat frames for PurusMat
Supplier:   PURUS
Mat frames for PurusMat
Supplier:  PURUS
Certificates
Mat frames for PurusMat
Mat frames for PurusMat
3 Options Available Below
Product Details & Documents
These frames are designed to be used with the Purus mats to create a system that is ideal for any critical area that does not allow direct adhesion to the flooring surface. The frames are constructed of a hard, rigid, PS base material combined with a non skid backing that holds the mats in place on almost any surface.
  • Protect floors from adhesive residue from the purus mats
  • Reduces the risk of slips and falls
  • Non-slip parts made from polymer coated polyester textile
  • Naturally antimicrobial, mould and mildew resistant
Product Family Options
Product Information
  • Colour
    For
    L×W
    Thickness
    Availability
    Price
  • White
    457×1143 mm
    508×1194 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
  • White
    660×1143 mm
    711×1194 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €
  • White
    914×1143 mm
    965×1194 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 290,00 €
    290,00 €
    1 * 1 ST
       290,00 €