Orders LinkContactUsLinkComponent
Spear sponges
Spear sponges
Disposable dissecting sponges
Catalog # HARV59-8422
Supplier:   HARVARD APPARATUS
Spear sponges
Catalog # HARV59-8422
Supplier:  HARVARD APPARATUS
Spear sponges
Spear sponges
Disposable dissecting sponges
Price
741,00 €
Log in for availability
1 * 200 ST
Product Details & Documents
These sterile cellulose sponges are designed for the removal of fluids during microsurgery.
  • Absorbs with only the tip immersed
  • Integral bendable plastic handle for added flexibility
  • Disposable for dissection use only
Suitable for spear applications.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Description:
    Disposable dissecting sponges