Order Entry
Belgium
Orders LinkContactUsLinkComponent
Waste Bin

Waste Bin

White, HDPE, 20 L, 300×290×400 mm
Catalog # LAMA1148
Supplier:   LAMAPLAST
Waste Bin
Catalog # LAMA1148
Supplier:  LAMAPLAST
Waste Bin
Waste Bin

White, HDPE, 20 L, 300×290×400 mm

Price
65,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

This dustbin with pedal and inside bucket is designed for ecological removal of garbages.

Specifications
  • Capacity:
    20 L
  • Colour:
    White
  • Material:
    HDPE
  • W×D×H:
    300×290×400 mm