Orders LinkContactUsLinkComponent
Hotplates
Hotplates
Supplier:   GESTIGKEIT HARRY
Hotplates
Supplier:  GESTIGKEIT HARRY
Certificates
18 Options Available Below
Product Details & Documents
Product Family Options
With performance control
Product Information
  • Heating area
    Temp.
    Heating capacity
    Type
    W×D×H
    Availability
    Price
  • 300×300 mm
    Max. 370 °C
    1800 W
    Hotplate, HD 0, aluminium alloy plate
    312×312×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 220,00 €
    1 220,00 €
    1 * 1 ST
       1 220,00 €
  • 350×350 mm
    Max. 370 °C
    2200 W
    Hotplate, HD 1, aluminium alloy plate
    358×358×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 350×500 mm
    Max. 370 °C
    2850 W
    Hotplate, HD 2, aluminium alloy plate
    364×514×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 590,00 €
    1 590,00 €
    1 * 1 ST
       1 590,00 €
  • 430×580 mm
    Max. 370 °C
    4000 W
    Hotplate, HD 3-230, aluminium alloy plate
    442×592×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 760,00 €
    1 760,00 €
    1 * 1 ST
       1 760,00 €
  • 430×580 mm
    Max. 370 °C
    4000 W
    Hotplate, HD 3-400, aluminium alloy plate
    442×592×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 760,00 €
    1 760,00 €
    1 * 1 ST
       1 760,00 €
  • 290×440 mm
    Max. 370 °C
    2400 W
    Hotplate, HE 1, aluminium alloy plate
    308×458×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 430,00 €
    1 430,00 €
    1 * 1 ST
       1 430,00 €
With performance control and thermostatic controller
Product Information
  • Heating area
    Temp.
    Heating capacity
    Type
    W×D×H
    Availability
    Price
  • 300×300 mm
    30...110 °C
    1150 W
    Hotplate, HT 01, aluminium alloy plate
    312×312×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 320,00 €
    1 320,00 €
    1 * 1 ST
       1 320,00 €
  • 300×300 mm
    50...300 °C
    1800 W
    Hotplate, HT 02, aluminium alloy plate
    312×312×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 320,00 €
    1 320,00 €
    1 * 1 ST
       1 320,00 €
  • 300×300 mm
    130...370 °C
    1800 W
    Hotplate, HT 03, aluminium alloy plate
    312×312×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 420,00 €
    1 420,00 €
    1 * 1 ST
       1 420,00 €
  • 350×350 mm
    50...300 °C
    2200 W
    Hotplate, HT 12, aluminium alloy plate
    358×358×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 450,00 €
    1 450,00 €
    1 * 1 ST
       1 450,00 €
  • 350×350 mm
    130...370 °C
    2200 W
    Hotplate, HT 13, aluminium alloy plate
    358×358×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 550,00 €
    1 550,00 €
    1 * 1 ST
       1 550,00 €
  • 350×500 mm
    30...110 °C
    1800 W
    Hotplate, HT 21, aluminium alloy plate
    364×514×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 680,00 €
    1 680,00 €
    1 * 1 ST
       1 680,00 €
  • 350×500 mm
    50...300 °C
    2850 W
    Hotplate, HT 22, aluminium alloy plate
    364×514×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 680,00 €
    1 680,00 €
    1 * 1 ST
       1 680,00 €
  • 350×500 mm
    130...370 °C
    2850 W
    Hotplate, HT 23, aluminium alloy plate
    364×514×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 770,00 €
    1 770,00 €
    1 * 1 ST
       1 770,00 €
  • 430×580 mm
    50...300 °C
    4000 W
    Hotplate, HT 32-230, aluminium alloy plate
    442×592×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 980,00 €
    1 980,00 €
    1 * 1 ST
       1 980,00 €
  • 430×580 mm
    50...300 °C
    4000 W
    Hotplate, HT 32-400, aluminium alloy plate
    442×592×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 980,00 €
    1 980,00 €
    1 * 1 ST
       1 980,00 €
  • 430×580 mm
    130...370 °C
    4000 W
    Hotplate, HT 33-230, aluminium alloy plate
    442×592×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 090,00 €
    2 090,00 €
    1 * 1 ST
       2 090,00 €
  • 430×580 mm
    130...370 °C
    4000 W
    Hotplate, HT 33-400, aluminium alloy plate
    442×592×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €