Orders LinkContactUsLinkComponent
Forceps, titanium
Forceps, titanium
Straight, sharp, 115 mm
Catalog # BOCH1910
Supplier:   BOCHEM
Forceps, titanium
Catalog # BOCH1910
Supplier:  BOCHEM
Forceps, titanium
Forceps, titanium
Straight, sharp, 115 mm
Price
64,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These titanium forceps are particularly suitable for applications where high corrosion-resistance, strength and low weight are important.
  • Lightweight, high strength
  • Corrosion and thermal resistance
  • Resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide
Specifications
  • Type:
    Straight, sharp
  • Length:
    115 mm
  • Seq:
    01