ContactUsLinkComponent
Detergent, liquid concentrate, Witonex-B-26
Detergent, liquid concentrate, Witonex-B-26
Catalog # WITG5.220.010
Supplier:  witeg Labortechnik
undefined
Detergent, liquid concentrate, Witonex-B-26
Catalog # WITG5.220.010
Supplier:  witeg Labortechnik
Specifications
  • Description:
    Witonex-B-26, liquid detergent, for rinsing washing machines
  • Pk:
    10 kg
Specifications
About this item
Liquid concentrate specially formulated to be low foaming for use with laboratory dishwashers.
  • For routine cleaning of objects and apparatus in medical, chemical and technical laboratories
  • Suitable for objects made of porcelain, glass, plastic, rubber and metals