Orders LinkContactUsLinkComponent
Container, with screw cap
Container, with screw cap
Supplier:   hünersdorff GmbH
Container, with screw cap
Supplier:  hünersdorff GmbH
Certificates
Container, with screw cap
Container, with screw cap
3 Options Available Below
Product Details & Documents
Containers with screw cap round, for varied applications, made of food-safe natural polypropylene.
  • Resistant to acids and chemicals
  • With tightly closing lid
  • Deep freezable
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Colour
    Availability
    Price
  • 250 ml
    104 mm
    50 mm
    Natural
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,70 €
    4,70 €
    1 * 1 ST
       4,70 €
  • 500 ml
    104 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 1000 ml
    125 mm
    126 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST