Orders LinkContactUsLinkComponent
Bottles, water sampling
Bottles, water sampling
Leverancier:   LP ITALIANA
Bottles, water sampling
Leverancier:  LP ITALIANA
Certificaten
Bottles, water sampling
Bottles, water sampling
2 Opties Hieronder Beschikbaar
Product Details & Documents
PP, translucent, square, with screw cap.
  • Tamper evident screw caps
  • Sterile
  • Moulded graduations
Bottles for microbiological analysis of chlorinated or non-chlorinated water. Sodium thiosulphate inhibits the effects of chlorine, bromide and ozone present in the water. Ideal for flat storage (fridges, etc).
Product Family Options
Productinformatie
  • Capacity
    Dosage
    Beschikbaarheid
    Price
  • 125 ml
    Undosed
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 350 ST
    1 * 350 ST - 560,00 €
    560,00 €
    1 * 350 ST
       560,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    LPIT299248
  • 125 ml
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 350 ST
    1 * 350 ST - 560,00 €
    560,00 €
    1 * 350 ST
       560,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    LPIT299148