Orders LinkContactUsLinkComponent
Storage Boxes for Vials
Storage Boxes for Vials
Supplier:   MACHEREY-NAGEL
Storage Boxes for Vials
Supplier:  MACHEREY-NAGEL
Certificates
Storage Boxes for Vials
Storage Boxes for Vials
5 Options Available Below
Product Details & Documents
Boxes are made from blue plastic with integrated or removable divider and transparent lid.
  • Allow a secure transportation of sample vials
  • Safe standing position in dividers designed for the respective diameter
  • Available for all 1,5 ml vials (standard volume), for crimp and screw neck vials N 13 and for headspace vials with screw neck N 18 or crimp neck N 20, respectively
  • Also available for EPA screw neck vials N 24 as well as for storage vials N 15
Ideal for space-saving storage in fridges and freezers, since the transparent lid prevents condensations on the closures and thus avoids a possible contamination in the cooling unit.
Documents
Product Family Options
Product Information
  • Compartments
    W×D×H ext.
    Availability
    Price
  • MANA702514
    Storage box for screw neck vials N 8, N 9, N 10 and crimp neck as well as snap ring vials N 11
    Storage box for screw neck vials N 8, N 9, N 10 and crimp neck as well as snap ring vials N 11
    81
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,60 €
    14,60 €
    1 * 1 ST
       14,60 €
  • MANA702515
    Storage box for crimp and screw neck vials N 13
    Storage box for crimp and screw neck vials N 13
    49
    130×130×50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,60 €
    15,60 €
    1 * 1 ST
       15,60 €
  • MANA702516
    Storage box for headspace screw neck vials N 18 and crimp neck vials N 20
    Storage box for headspace screw neck vials N 18 and crimp neck vials N 20
    25
    130×130×80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • MANA702517
    Storage box for EPA screw neck vials N 24 (20 ml, 30 ml, 40 m)
    Storage box for EPA screw neck vials N 24 (20 ml, 30 ml, 40 m)
    16
    130×130×102 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,50 €
    34,50 €
    1 * 1 ST
       34,50 €
  • MANA702518
    Storage box for screw neck vials N 15 (sample storage)
    Storage box for screw neck vials N 15 (sample storage)
    36
    130×130×80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,60 €
    19,60 €
    1 * 1 ST
       19,60 €