Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boxes
Storage Boxes
Accessories for Laboratory Containers
Accessories for Laboratory Containers
Accessories for Laboratory Containers
Catalog # SEMA2115
Supplier:
SEMADENI
Expand image
Accessories for Laboratory Containers
Catalog # SEMA2115
Supplier:
SEMADENI
Specifications
Description:
Lid for storage and transport container 220×120 mm
Seq:
01
Pk:
1
Specifications
About this item
Product Details
Product Details