Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type

Sedimentation cones, Imhoff type

Capacity 1000 ml
Catalog # BIBBCRF530
Supplier:   SciLabware
Sedimentation cones, Imhoff type
Catalog # BIBBCRF530
Supplier:  SciLabware
Sedimentation cones, Imhoff type
Sedimentation cones, Imhoff type

Capacity 1000 ml

Price
273,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass.

  • Graduated up to 40 ml and with capacity mark at 1000 ml

For determining the amount of sediment in sewage or industrial waste.

Specifications
  • Description:
    Capacity 1000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.