Orders LinkContactUsLinkComponent
Conductivity cells, Radiometer Analytical
Conductivity cells, Radiometer Analytical
Supplier:   Hach
Conductivity cells, Radiometer Analytical
Supplier:  Hach
Certificates
Conductivity cells, Radiometer Analytical
Conductivity cells, Radiometer Analytical
11 Options Available Below
Product Details & Documents
Range of conductivity cells for a wide variety of applications.
  • Two-pole cells: traditional design based on two plates of platinum; ideal for routine measurement of conductivity and for use with a sample changer due to the easy rinsing
  • three-pole cells: three platinum rings which facilitate optimal shielding during measurement
  • four-pole cells: four platinum rings ensure accurate results over several decades of conductivity with a single cell using just one calibration
  • four-pole cells are particularly recommended when performing high conductivity measurements
Product Family Options
Product Information
  • Type
    Cell constant
    Material
    Ø×L
    Connection
    For
    Availability
    Price
  • HACHB60E100
    Conductivity cell, 2-pole
    Conductivity cell, 2-pole
    XE100, 2-poles
    1,0 cm⁻¹
    Glass
    12×120 mm
    Screw cap
    Aqueous and non aqueous media
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 515,00 €
    515,00 €
    1 * 1 ST
       515,00 €
  • HACHE61M008
    Conductivity cell, 2-pole
    Conductivity cell, 2-pole
    CDC241-9, 2-pole
    1,0 cm⁻¹
    Glass
    12×103 mm
    Screw cap
    Viscous titrations
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 517,00 €
    517,00 €
    1 * 1 ST
       517,00 €
  • HACHE61M011
    Conductivity cell, 2-pole
    Conductivity cell, 2-pole
    CDC267, 2-pole
    0,1 cm⁻¹
    Glass
    12×103 mm
    Screw cap
    Highly resistant media
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HACHE61M013
    Conductivity cell, 2-pole
    Conductivity cell, 2-pole
    CDC745-9, 2-pole
    1,0 cm⁻¹
    Epoxy
    12×103 mm
    Screw cap
    Field use
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 466,00 €
    466,00 €
    1 * 1 ST
       466,00 €
  • HACHE61M014
    Conductivity cell, 2-pole
    Conductivity cell, 2-pole
    CDC749, 2-poles
    1,7 cm⁻¹
    Glass
    4×103 mm
    MAB6 plug head, fixed cable
    Micro samples in glass tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 878,00 €
    878,00 €
    1 * 1 ST
       878,00 €
  • HACHB15B001
    Conductivity cell, 2-pole, with temp. sensor
    Conductivity cell, 2-pole, with temp. sensor
    CDC641T, 2-pole
    0,85 cm⁻¹
    Glass
    12×103 mm
    MAB6 plug head, fixed cable
    Aqueous and non aqueous media
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 769,00 €
    769,00 €
    1 * 1 ST
       769,00 €
  • HACHE61M012
    Conductivity cell, 2-pole, with temp. sensor
    Conductivity cell, 2-pole, with temp. sensor
    CDC741T, 2-pole
    1,0 cm⁻¹
    Glass
    12×103 mm
    MAB6 plug head, fixed cable
    Use with sample changer
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 921,00 €
    921,00 €
    1 * 1 ST
       921,00 €
  • HACHE61M022
    Conductivity cell, 2-pole, with temp. sensor
    Conductivity cell, 2-pole, with temp. sensor
    CDC641T-6, 2-pole
    0,85 cm⁻¹
    Glass
    12×103 mm
    RJ45 plug head, fixed cable
    Aqueous and non aqueous media
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 900,00 €
    900,00 €
    1 * 1 ST
       900,00 €
  • HACHE61M009
    Conductivity cell, 4-pole, with temp. sensor
    Conductivity cell, 4-pole, with temp. sensor
    CDC511T, 4-pole
    1,0 cm⁻¹
    Polymer fibres
    12×114 mm
    MAB6 plug head, fixed cable
    Pure water
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 640,00 €
    1 640,00 €
    1 * 1 ST
       1 640,00 €
  • HACHE61M010
    Conductivity cell, 4-pole, with temp. sensor
    Conductivity cell, 4-pole, with temp. sensor
    CDC566T, 4-pole
    1,0 cm⁻¹
    Epoxy
    12×103 mm
    MAB6 plug head, fixed cable
    General purpose, field use
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HACHE61M015
    Conductivity cell, 4-pole, with temp. sensor
    Conductivity cell, 4-pole, with temp. sensor
    CDC866T, 4-pole
    1,0 cm⁻¹
    Epoxy
    12×103 mm
    MAB6 plug head, fixed cable
    General purpose, field use
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €