Orders LinkContactUsLinkComponent
Laboratory reactor system
Laboratory reactor system, 220 °C, 6 bar, complete, includes 2 l glass vessel, 316 stainless steel lid fitted with stirrer head and heating jacket, pressure gauge, excess pressure valve, thermometer pocket, B29 adaptor with pressure tight cap, inlet with plug valve, tripod and a safety cage
Catalog # JUCH10101.12
Laboratory reactor system
Catalog # JUCH10101.12
Laboratory reactor system
Laboratory reactor system
Laboratory reactor system, 220 °C, 6 bar, complete, includes 2 l glass vessel, 316 stainless steel lid fitted with stirrer head and heating jacket, pressure gauge, excess pressure valve, thermometer pocket, B29 adaptor with pressure tight cap, inlet with plug valve, tripod and a safety cage
Price
26 100,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Laboratory reactor system, 220 °C, 6 bar, complete, includes 2 l glass vessel, 316 stainless steel lid fitted with stirrer head and heating jacket, pressure gauge, excess pressure valve, thermometer pocket, B29 adaptor with pressure tight cap, inlet with plug valve, tripod and a safety cage
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lid, stainless steel, without fittings
    Lid, stainless steel, without fittings
    JUCH10101.20
    Log in for availability
    1 * 1 ST - 6 240,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare bolt springs, each
    Spare bolt springs, each
    JUCH10101.06
    Log in for availability
    1 * 1 ST - 44,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare compression ring
    Spare compression ring
    JUCH10101.05
    Log in for availability
    1 * 1 ST - 1 630,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare ‘O’ ring
    Spare ‘O’ ring
    JUCH10101.04
    Log in for availability
    1 * 1 ST - 271,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare pressure equalising spring
    Spare pressure equalising spring
    JUCH10101.03
    Log in for availability
    1 * 1 ST - 122,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Rear wall, additional, wire netting
    Rear wall, additional, wire netting
    JUCH10101.02
    Log in for availability
    1 * 1 ST - 681,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Safety cage, three sided
    Safety cage, three sided
    JUCH10101.01
    Log in for availability
    1 * 1 ST - 3 700,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass vessel body with jacket, 2 l
    Glass vessel body with jacket, 2 l
    JUCH10101.00
    Log in for availability
    1 * 1 ST - 6 040,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Gaskets, PTFE, for conical joint adaptors B29 with caps, Ø 36×30 mm
    Gaskets, PTFE, for conical joint adaptors B29 with caps, Ø 36×30 mm
    JUCH20336.31
    Log in for availability
    1 * 1 ST - 13,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Double joint, stainless steel
    Double joint, stainless steel
    JUCH20440.11
    Log in for availability
    1 * 1 ST