Order Entry
Belgium
ContactUsLinkComponent
 
Wall clock, radio controlled, 5602GL
  MOIN5602GL
 :  MOINEAU
undefined
Wall clock, radio controlled, 5602GL
  MOIN5602GL
 :  MOINEAU

 

  • Ø:
    300 mm
  • Colour:
    Black
  • Description:
    Wall clock, radio controlled, 5602GL, weight 750 g
  • Pk:
    1

 

 

This radio-controlled wall clock is readable from a distance of up to 20 m.

  • ABS / mineral glass
  • Low battery indicator