Order Entry
Belgium
ContactUsLinkComponent
UPLC columns, ACQUITY UPLC™ CSH phenyl-hexyl
UPLC columns, ACQUITY UPLC™ CSH phenyl-hexyl
Catalog # WATE186005406
Supplier:  WATERS
UPLC columns, ACQUITY UPLC™ CSH phenyl-hexyl
Catalog # WATE186005406
Supplier:  WATERS

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Particle Size:
    1,7 µm
  • Pore size:
    130 Å
  • Length:
    50 mm
  • Phase:
    Phenyl-hexyl
  • Pk:
    1

Specifications

About this item

These columns provides complementary selectivity to straight-chain-alkyl phases, particularly for polyaromatic compounds. Built on Charged Surface Hybrid (CSH) particle platform.