ContactUsLinkComponent
UPLC columns, ACQUITY UPLC™ BEH Shield RP 18
 :  WATERS
UPLC columns, ACQUITY UPLC™ BEH Shield RP 18
 :  WATERS
  • Catalog No:
  • WATE186002853
  • WATE186002854
  • Carbon load:
  • 17%
  • 17%
  • Endcapped:
  • Yes, Yes
  • Yes, Yes
  • pH range:
  • 2 - 11
  • 2 - 11
  • Surface area:
  • 185 m²/g
  • 185 m²/g
  • USP code:
  • L1, L1
  • L1, L1
Frequently Bought Together