UPLC columns, ACQUITY UPLC™ BEH phenyl
Fournisseur: WATERS
Certificats
À propos de cet article
These columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
- Chemically stable phenyl sorbent
- Ultra-low column bleed, long column lifetimes and excellent peak shape
Spécifications
- Catalog No:
- WATE186002884
- WATE186003911
- WATE186002885
- WATE186003375
- Carbon load:
- 15%
- 15%
- 15%
- 15%
- Endcapped:
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- pH range:
- 1 - 12
- 1 - 12
- 1 - 12
- 1 - 12
- Surface area:
- 185 m²/g
- 185 m²/g
- 185 m²/g
- 185 m²/g
- USP code:
- L11, L11
- L11, L11
- L11, L11
- L11, L11
Spécifications
Frequently Bought Together