Affinity chromatography column, MAbPac™ Protein A
HYPE082539
- Column Format:Analytical
- Ø int.:4,00 mm
- Particle Size:12 µm
- Max. pressure:1000 psi
- Length:35 mm
- Seq:01
- pH range:2,5 - 7,5
- Temperature limit:30 °C
- Column hardware:PEEK
- Packing material:Spherical, non-porous polymer
- Pk:1
For fast, accurate titer analysis of monoclonal antibodies (MAbs) in harvest cell cultures. This column provides high throughput and accurate analyses through a combination of low backpressure and high efficiency. Its unique format makes automation of loading, binding, elution and collection using biocompatible systems quick and easy. A non-porous, polymeric resin with a hydrophobic divinylbenzene core and a hydrophilic surface optimised for affinity separation makes this column rugged and reproducible.
- Complete cycle time, including equilibration, of two minutes
- Linear over a wide concentration range (0,025 to 5 mg/ml) without carryover
- Low column pressure of less than 250 psi at 2 ml/min