Orders LinkContactUsLinkComponent
Measuring scoops, PP
Measuring scoops, PP
Supplier:   SEMADENI
Measuring scoops, PP
Supplier:  SEMADENI
Certificates
Measuring scoops, PP
Measuring scoops, PP
7 Options Available Below
Product Details & Documents
PP, white. Shatterproof measuring scoops with good chemical resistance.
  • With flat, bevelled end and reinforced handle
  • Heat resistant up to 130 °C, sterilisable
  • Volume is visible on the handle of each scoop
The scoops are also food contact approved and can therefore be used for food manufacturing or processing applications.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Length
    Width
    Colour
    Availability
    Price
  • 5 ml
    82 mm
    25 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,90 €
    1,90 €
    1 * 1 ST
       1,90 €
  • 10 ml
    100 mm
    35 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,40 €
    2,40 €
    1 * 1 ST
       2,40 €
  • 25 ml
    135 mm
    45 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,70 €
    2,70 €
    1 * 1 ST
       2,70 €
  • 50 ml
    160 mm
    55 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,50 €
    3,50 €
    1 * 1 ST
       3,50 €
  • 100 ml
    200 mm
    65 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,10 €
    5,10 €
    1 * 1 ST
       5,10 €
  • 250 ml
    260 mm
    90 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,70 €
    11,70 €
    1 * 1 ST
       11,70 €
  • 1000 ml
    385 mm
    150 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,80 €
    20,80 €
    1 * 1 ST
       20,80 €
This product has been blocked by your organization. Please contact your purchasing department for more information