Orders LinkContactUsLinkComponent
Safety baskets
Safety baskets
Supplier:   SEMADENI
Safety baskets
Supplier:  SEMADENI
Certificates
Safety baskets
3 Options Available Below
Product Details & Documents
PP, coloured.
  • Anti-static
  • Space-saving, stackable
  • Good chemical and temperature resistance
The antistatic safety baskets with perforated walls and base comes with collapsible plastic mounting bracket with lock in vertical position.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • SEMA2699
    Basket with handle, PP, yellow
    Basket with handle, PP, yellow
    430×310×220 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,00 €
    49,00 €
    1 * 1 ST
       49,00 €
  • SEMA4953
    Basket with handle, PP, blue
    Basket with handle, PP, blue
    430×310×220 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,80 €
    48,80 €
    1 * 1 ST
       48,80 €
  • SEMA4954
    Basket with handle, PP, red
    Basket with handle, PP, red
    430×310×220 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,40 €
    46,40 €
    1 * 1 ST
       46,40 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.