Orders LinkContactUsLinkComponent
Cell viability assay kits
Supplier:   Biotium
Cell viability assay kits
Supplier:  Biotium
Certificates
Cell viability assay kits
4 Options Available Below
Product Details & Documents
MTT, XTT, and resazurin are reduced by mitochondrial metabolic activity to yield coloured or fluorescent products, and thus are useful for and assaying cell viability and quantitating cell number. MTT and XTT are reduced to coloured formazin salts that can be measured by absorbance. MTT generates an insoluble formazin salt, requiring cell lysis before the absorbance can be measured, while XTT does not require cell lysis for measurement. Resazurin is a non-fluorescent blue dye that is reduced to the pink fluorescent compound resorufin, which can be measured by fluorescence or absorbance.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 30007.
    XTT Cell Viability Assay kit, 1000 assays
    XTT Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 308,00 €
    308,00 €
    1 * 1 KIT
       308,00 €
  • 30006.
    MTT Cell Viability Assay kit, 1000 assays
    MTT Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 256,00 €
    256,00 €
    1 * 1 KIT
       256,00 €
  • 30025-1
    Resazurin Cell Viability Assay kit, 2500 assays (25 ml)
    Resazurin Cell Viability Assay kit, 2500 assays (25 ml)
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 219,00 €
    219,00 €
    1 * 1 KIT
       219,00 €
  • 30025-2
    Resazurin Cell Viability Assay kit, 10 000 assays (100 ml)
    Resazurin Cell Viability Assay kit, 10 000 assays (100 ml)
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 518,00 €
    518,00 €
    1 * 1 KIT
       518,00 €