ContactUsLinkComponent
Versene
Versene
Catalog # L0630-100
Supplier:  Biowest
CAS Number:  
undefined
Versene
Catalog # L0630-100
Supplier:  Biowest
CAS Number:  
Specifications
  • Pk:
    100 ml
  • Description:
    Versene
Specifications
About this item
Versene can be used instead of trypsin. It is a chelating agent that disperses the cells by cutting the cytoplasmic bridges between them. Versene has the advantage of being able to withstand autoclave temperature, which is a guarantee of sterility. In addition, it is less aggressive cells than trypsin, which is useful for studies on cell growth.